BAF171310018
SP Bel-Art® Pipette Rinser for Cleanware Pipette Rinsing System
diam. × H 25.2 cm (9.9 in.) × 64 cm (25.25 in.)
All Photos(3)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
Recommended Products
feature
to hold For 46 cm (18") pipettes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F17131-0018
diam. × H
25.2 cm (9.9 in.) × 64 cm (25.25 in.)
General description
The Cleanware Pipette Rinsing System for pipette cleaning and storage has an effective, reliable design that ensures safe, efficient operation. Available in a complete system or in individual components.
Features and Benefits
- Pipette Rinsers provides a simple, automatic method for rinsing large numbers of pipettes at one time
- High level water entry provides an air gap between the maximum water level and the end of the water inlet tube ensuring safe and efficient operation
- “Tube within a tube” siphoning system assures rapid, complete and automatic siphoning and refilling over a wide range of inlet water pressures and flow rates
- Minimizes the danger of siphon stalling at certain water inflow rates and prevents any possible backflow into the potable water supply in the event of a pressure interruption
- Leak-proof, polyethylene Pipette Jars store and soak pipettes and will withstand all common cleaning solutions at room temperature, including sulfuric acid/dichromate
- Pipette Baskets, filled with pipettes, fit into the corresponding size jar; cut-out hand grips for easy handling; gusher holes provide quick drainage when lifted from a soaking jar; foam disk on the bottom cushions pipette tips to prevent breakage
- Includes 30.5cm (12”) long piece of 2.5cm (1”) I.D. drain tube and 1.5M (5ft) of 12.5mm (½”) I.D. flexible water inlet tube
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP