Skip to Content
Merck
All Photos(3)

Key Documents

531111

Sigma-Aldrich

N-Hydroxy-5-norbornene-2,3-dicarboximide perfluoro-1-butanesulfonate

electronic grade, ≥99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8F9NO5S
CAS Number:
Molecular Weight:
461.26
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

description

Total acid <2 × 10-3 mol/kg
Total base <2 × 10-3 mol/kg

Assay

≥99% trace metals basis

mp

55-59 °C (lit.)

solubility

γ-butyrolactone: 50%
PGMEA: >50%
ethyl lactate: 50%

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)ON1C(=O)C2C3CC(C=C3)C2C1=O

InChI

1S/C13H8F9NO5S/c14-10(15,12(18,19)20)11(16,17)13(21,22)29(26,27)28-23-8(24)6-4-1-2-5(3-4)7(6)9(23)25/h1-2,4-7H,3H2/t4-,5+,6?,7?

InChI key

PNXSDOXXIOPXPY-DPTVFECHSA-N

Application

Cationic photoinitiator. Nonionic photoacid generator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service