Merck
  • Insecticide-mediated apparent displacement between two invasive species of leafminer fly.
Insecticide-mediated apparent displacement between two invasive species of leafminer fly.
PloS one (2012-06-05)
Yulin Gao, Stuart R Reitz, Qingbo Wei, Wenyan Yu, Zhongren Lei
ABSTRACT
Closely related invasive species may often displace one another, but it is often difficult to determine mechanisms because of the historical nature of these events. The leafmining flies Liriomyza sativae and Liriomyza trifolii have become serious invasive agricultural pests throughout the world. Where both species have invaded the same region, one predominates over the other. Although L. sativae invaded Hainan Island of China first, it recently has been displaced by the newly invasive L. trifolii. We hypothesized that differential susceptibilities to insecticides could be causing this demographic shift. Avermectin and cyromazine are the most commonly used insecticides to manage leafminers, with laboratory bioassays demonstrating that L. trifolii is significantly less susceptible to these key insecticides than is L. sativae. In trials where similar numbers of larvae of both species infested plants, which subsequently were treated with the insecticides, the eclosing adults were predominately L. trifolii, yet similar numbers of adults of both species eclosed from control plants. The species composition was then surveyed in two regions where L. trifolii has just begun to invade and both species are still common. In field trials, both species occurred in similar proportions before insecticide treatments began. Following applications of avermectin and cyromazine, almost all eclosing adults were L. trifolii in those treatment plots. In control plots, similar numbers of adults of the two species eclosed, lending further credence to the hypothesis that differential insecticide susceptibilities could be driving the ongoing displacement of L. sativae by L. trifolii. Our results show that differential insecticide susceptibility can lead to rapid shifts in the demographics of pest complexes. Thus, successful pest management requires the identification of pest species to understand the outcome of insecticide applications. These results further demonstrate the importance of considering anthropogenic factors in the outcome of interspecific interactions.
Product Description
Supelco
Cyromazin, PESTANAL®, analytical standard
Sigma-Aldrich
N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine, 97%