Merck
  • Ultrasound-promoted catalyst-free one-pot four component synthesis of 2H-indazolo[2,1-b]phthalazine-triones in neutral ionic liquid 1-butyl-3-methylimidazolium bromide.
Ultrasound-promoted catalyst-free one-pot four component synthesis of 2H-indazolo[2,1-b]phthalazine-triones in neutral ionic liquid 1-butyl-3-methylimidazolium bromide.
Ultrasonics sonochemistry (2011-08-27)
Mohsen Shekouhy, Alireza Hasaninejad
ABSTRACT
A catalyst-free one-pot four component methodology for the synthesis of 2H-indazolo[2,1-b]phthalazine-triones under ultrasonic irradiation at room temperature using 1-butyl-3-methylimidazolium bromide, [Bmim]Br, as a neutral reaction medium is described. A broad range of structurally diverse aldehydes (aromatic aldehydes bearing electron withdrawing and/or electron releasing groups as well as heteroaromatic aldehydes) were applied successfully, and corresponding products were obtained in good to excellent yields without any byproduct.
Sigma-Aldrich
Phthalazine, 98%