- Phosphorylation of glyceric acid in aqueous solution using trimetaphosphate.
Phosphorylation of glyceric acid in aqueous solution using trimetaphosphate.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (1996-02-01)
V Kolb, L E Orgel
PMID11536746
ABSTRACT
The phosphorylation of glyceric acid is an interesting prebiotic reaction because it converts a simple, potentially prebiotic organic molecule into phosphate derivatives that are central to carbohydrate metabolism. We find that 0.05 M glyceric acid in the presence of 0.5 M trimetaphosphate in alkaline solution gives a mixture of 2- and 3-phosphoglyceric acids in combined yields of up to 40%.