Z358290
Nalgene® staining box
L × W × H 12.5 cm × 12.5 cm × 5 cm
Synonym(s):
electrophoresis tray, gel staining box
All Photos(1)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.22
Recommended Products
material
(PCTG)
packaging
pack of 2 ea
manufacturer/tradename
Nalgene® 5705-1010
L × W × H
12.5 cm × 12.5 cm × 5 cm
Related Categories
General description
Constructed of PCTG [poly(1,4-cyclohexylene dimethylene terephthalate)glycol] with a polypropylene copolymer cover and PDPE plug. Designed for staining and destaining electrophoresis gels. Resists temperatures from −70 to +70°C and is resistant to acids, bases, and some organic solvents. Plug allows draining without handling gels. Stackable.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)