Skip to Content
Merck
All Photos(1)

Key Documents

SML1451

Sigma-Aldrich

MI192 hydrochloride

≥98% (HPLC)

Synonym(s):

N-(2-Aminophenyl)-4-[(3,4-dihydro-4-methylene-1-oxo-2(1H)-isoquinolinyl)methyl]-benzamide hydrochloride, MI-192 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21N3O2 · xHCl
CAS Number:
Molecular Weight:
383.44 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C=C(CN1CC2=CC=C(C(NC3=CC=CC=C3N)=O)C=C2)C4=CC=CC=C4C1=O

InChI

1S/C24H21N3O2/c1-16-14-27(24(29)20-7-3-2-6-19(16)20)15-17-10-12-18(13-11-17)23(28)26-22-9-5-4-8-21(22)25/h2-13H,1,14-15,25H2,(H,26,28)

InChI key

GTLTXEIKQVWSRF-UHFFFAOYSA-N

Biochem/physiol Actions

MI192 is a potent and selective histone deacetylase 3 (HDAc 3) inhibitor that inhibits IL-6 production in peripheral blood mononuclear cells (PBMCs) from rheumatoid arthritis (RA) but not healthy PBMCs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service