Skip to Content
Merck
All Photos(1)

Key Documents

EMU078351

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Spag9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACGCACTGCATCAGAGACACACTGAGATGATCCATAATTATATGGAACACTTAGAAAGAACCAAACTTCATCAGCTCTCAGGGAGTGATCAACTAGAGGCTACAGCTCATAGTAGAATTAGAAAAGAACGTCCTATATCATTAGGGATTTTCCCTCTACCTGCTGGAGATGGATTGCTTACACCTGACACTCAGAAAGGAGGCGAGACCCCAGGATCAGAGCAATGGAAATTTCAAGAATTAAGTCAACCATGTTCTCATACCAGCCTGAAGGATGAACTTTCTGATATTAGTCAAGGTGGATCTAAAGCTACCACTCCAGCTTCAACAGCAAATTCAGATGTATCAGCAATTCCTCCTGATACTCCGTCAAAGGAAGATAATGAAGGATTTGTAAAAGGCACAGATACATCAAATAAGTCAGAGATAAGCAAACACATAGAAGTCCAGGTTGCCCA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Li et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(7), 6949-6954 (2014-04-18)
Sperm-associated antigen 9 (SPAG9) was recently reported to be overexpressed in several cancers and associated with the malignant behavior of cancer cells. However, the expression pattern of SPAG9 and its clinical significance in human prostate cancer have not been reported.
Zhi-Feng Miao et al.
Virchows Archiv : an international journal of pathology, 467(5), 525-533 (2015-08-22)
Sperm associated antigen 9 (SPAG9) protein has been found to play an important role in cancer progression but the involved mechanisms are still obscure. Its clinical significance in human gastric cancers remains unexplored. In the present study, SPAG9 expression was
Feifei Chen et al.
Oncology reports, 32(6), 2533-2540 (2014-10-14)
Sperm-associated antigen 9 (SPAG9) is a recently characterized oncoprotein involved in the progression of several human malignancies. To elucidate the role of SPAG9 in the development of human prostate cancer (PCa), tissue microarray (TMA) and immunohistochemistry were used to detect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service