Skip to Content
Merck
All Photos(1)

Key Documents

8.04788

Sigma-Aldrich

1-Iodopropane

(stabilised with sodium carbonate) for synthesis

Synonym(s):

1-Iodopropane, Propyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7I
CAS Number:
Molecular Weight:
169.99
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-460-2
NACRES:
NA.22

vapor pressure

57 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

103 °C/1013 mbar

mp

-98 °C

transition temp

flash point 44 °C

solubility

1.1 g/L

density

1.75 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

IC(C)C

InChI

1S/C3H7I/c1-3(2)4/h3H,1-2H3

InChI key

FMKOJHQHASLBPH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.740 - 1.750
Identity (IR): passes test

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service