Skip to Content
Merck
All Photos(1)

Key Documents

5.31552

Sigma-Aldrich

Argonaute-2 Inhibitor, BCI-137

Synonym(s):

Argonaute-2 Inhibitor, BCI-137, 2-(2,3-Dioxo-1,2,3,4-tetrahydroquinoxaline-6-sulfonamido) propanoic acid, N-((2,3-Dihydroxy-6-quinoxalinyl)sulfonyl)alanine, Ago2 Inhibitor, BCI137

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N3O6S
CAS Number:
Molecular Weight:
313.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

126 μM Kd

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

gray-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CC(C(=O)O)NS(=O)(=O)C1=CC2=C(C=C1)NC(=O)C(=O)N2

General description

A cell-permeable dioxotetrahydroquinoxaline compound that mimics uridine monophosphate for affinity interaction at the Argonaute-2 (Ago2) Mid domain (KD = 126 µM) and effectively inhibits Ago2 cellular miRNA loading in acute promyelocytic leukemia (APL) NB4 cultures (% inhibition by RIP = 79%/miR-107, 73%/let-7a, 73%/miR-223, 68%/miR-26a, 53%/miR-60a; 100 nM for 48 h) without affecting cellular Ago2 stability. Consistent with the involvement of Ago2 in heterochromatic miR-155HG silencing, BCI-137 is reported to decrease miR-155HG promoter region-associated Ago2 with a concomitant acetylation induction of the promoter region-associated histone H4 (70% reduced Ago2 association and 300% enhanced H4 acetylation, respectively, by ChIP; 10 µM). Likewise, Ago2 inhibition by BCI-137 is shown to boost Retinoic Acid-(RA, Cat. no. 554720) induced NB4 granulocyte differentiation as a result of greatly upregulated expression of the differentiation-related miRNAs (140% and 255% increased miR-26a and miR-223 level, respectively, compared to 100 nM RA stimulation alone for 96 h) without affecting NB4 proliferation or viability (up to 10 µM and 96 h with or without 100 nM RA stimulation).
A cell-permeable, non-toxic dioxotetrahydroquinoxaline compound that mimics uridine and directly and reversibly interacts with the microRNA (miRNA) binding domain of Argonaute-2 (Ago2; KD = 126 µM) and significantly inhibits binding of miR-20a, miR-26a, miR-107, miR-223, and let-7a to Ago2. However, it does not affect Ago2 stability in cells. Displaces Ago2 from the miR-155 host gene promoter region (70% reduction in Ago2 association) and increases histone H4 acetylation (~300%) in NB4 leukemia cell line. Promotes retinoic acid-induced differentiation of NB4 cells to granulocytes.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Argonaute-2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Masciarelli, S., et al. 2014. ACS Chem. Biol.9, 1674.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service