Skip to Content
Merck
All Photos(1)

Key Documents

800740P

Avanti

N-16:0 L-Serine

N-palmitoyl L-serine, powder

Synonym(s):

1-hexadecanoyl-L-serine; (S)-3-hydroxy-2-hexadecanamidopropanoate; (S)-3-hydroxy-2-palmitamidopropanoate; 110831

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H37NO4
CAS Number:
Molecular Weight:
343.50
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (800740P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800740P

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].N([C@@H](CO)C(=O)[O-])C(=O)CCCCCCCCCCCCCCC

InChI

1S/C19H37NO4.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(22)20-17(16-21)19(23)24;/h17,21H,2-16H2,1H3,(H,20,22)(H,23,24);/q;+1/p-1/t17-;/m0./s1

InChI key

XKMKJRANXQGSEK-LMOVPXPDSA-M

Biochem/physiol Actions

N-16:0 L-Serine serves as an antagonist for lysophosphatidic acid (LPA) receptor. It prevents platelet aggregation and chloride conductance induced by LPA.

Packaging

5 mL Amber Glass Screw Cap Vial (800740P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis of phosphatidylserine-exposing red blood cells by secretory phospholipase A2 generates lysophosphatidic acid and results in vascular dysfunction
Neidlinger NA, et al.
The Journal of biological chemistry, 281(2), 775-781 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service