Skip to Content
Merck
All Photos(1)

Key Documents

PZ0335

Sigma-Aldrich

CP724714

≥98% (HPLC)

Synonym(s):

(E)-2-Methoxy-N-[3-[4-[[3-methyl-4-[(6-methylpyridin-3-yl)oxy]phenyl]amino]quinazolin-6-yl]allyl]ethanamide, 2-Methoxy-N-[(2E)-3-[4-[[3-methyl-4-[(6-methyl-3-pyridinyl)oxy]phenyl]amino]-6-quinazolinyl]-2-propen-1-yl]-acetamide, CP-724,714, CP-724714

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27N5O3
CAS Number:
Molecular Weight:
469.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

COCC(NC/C=C/C1=CC=C(N=CN=C2NC3=CC=C(OC4=CC=C(C)N=C4)C(C)=C3)C2=C1)=O

InChI

1S/C27H27N5O3/c1-18-13-21(8-11-25(18)35-22-9-6-19(2)29-15-22)32-27-23-14-20(7-10-24(23)30-17-31-27)5-4-12-28-26(33)16-34-3/h4-11,13-15,17H,12,16H2,1-3H3,(H,28,33)(H,30,31,32)/b5-4+

InChI key

LLVZBTWPGQVVLW-SNAWJCMRSA-N

Gene Information

human ... ERBB2(2064)

Biochem/physiol Actions

CP724714 is a potent and selective inhibitor of HER2/ErbB2 with an IC50 value of 10 nM. CP724714 is >640-fold selectivity against EGFR, and >1000-fold selective against InsR, IRG-1R, PDGFR, VEGFR2, Abl, Src, c-Met, JNK-2, JNK-3, ZAP-70, CDK-2 and CDK-5 in in vitro kinase assays. CP724714 had antiproliferative effects in a panel of human breast cancer cell lines.

Other Notes

CP-724714 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the CP-724714 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service