Skip to Content
Merck
All Photos(1)

Key Documents

M6057

Sigma-Aldrich

Anti-Matrix Metalloproteinase-12, Propeptide Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 54 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:3,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP12(4321)

General description

Matrix metalloproteinase-12 (MMP-12) is also called as macrophage metalloelastase and is secreted by inflammatory macrophages. It is synthesized as a zymogen and then activated by cleaving the propeptide domain. The MMP-12 gene is localized on human chromosome 11.

Specificity

The antibody recognizes only the latent form of MMP-12 and not the active forms. May be used to distinguish the zymogen and the activated enzyme.

Immunogen

synthetic peptide corresponding to the propeptide region of human matrix metalloproteinase-12 (macrophage elastase).

Biochem/physiol Actions

Matrix metalloproteinase-12 (MMP-12) is involved in breaking down proteoglycans, collagen type IV and laminin. Its main substrate is elastin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Song et al.
BMC cardiovascular disorders, 13, 34-34 (2013-05-07)
Aortic dissection(AD) is an acute process of large blood vessels characterized by dangerous pathogenic conditions and high disability and high mortality. The pathogenesis of AD remains debated. Matrix metalloproteinase-12 (MMP-12) participates in many pathological processes such as abdominal aortic aneurysm
Matthew Traylor et al.
PLoS genetics, 10(7), e1004469-e1004469 (2014-08-01)
Genome-wide association studies (GWAS) have begun to identify the common genetic component to ischaemic stroke (IS). However, IS has considerable phenotypic heterogeneity. Where clinical covariates explain a large fraction of disease risk, covariate informed designs can increase power to detect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service