Skip to Content
Merck
All Photos(1)

Key Documents

43424

Sigma-Aldrich

N-Isovalerylglycine

≥97.0% (GC)

Synonym(s):

N-(3-Methyl-1-oxobutyl)glycine, N-(3-Methylbutanoyl)glycine, [(3-Methylbutanoyl)amino]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO3
CAS Number:
Molecular Weight:
159.18
Beilstein:
1766622
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

N-Isovalerylglycine, ≥97.0% (GC)

Quality Level

Assay

≥97.0% (GC)

form

powder or crystals

color

white to off-white

mp

87-90 °C

format

neat

InChI

1S/C7H13NO3/c1-5(2)3-6(9)8-4-7(10)11/h5H,3-4H2,1-2H3,(H,8,9)(H,10,11)

InChI key

ZRQXMKMBBMNNQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Isovalerylglycine is a byproduct of the catabolism of the amino acid leucine. Accumulation of isovalerylglycine occurs in Isovaleric Acidemia (IVA), an autosomal recessive disorder caused by mutations in the isovaleryl-CoA dehydrogenase gene. The deficiency of this enzyme in the metabolism of leucine leads to the accumulation of a series of isovaleryl-CoA metabolites, such as isovalerylglycine (IVG).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D G Hine et al.
Pediatric research, 20(3), 222-226 (1986-03-01)
A stable isotope dilution method was developed to measure accurately small amounts of isovalerylglycine in amniotic fluid and urine for the prenatal diagnosis of isovaleric acidemia. [4,4,4-D3]Isovalerylglycine was synthesized and used as an internal standard. Samples were extracted, methylated, and
Yosuke Shigematsu et al.
Clinica chimica acta; international journal of clinical chemistry, 386(1-2), 82-86 (2007-09-14)
Recent neonatal screening for isovaleric acidemia by tandem mass spectrometry based on dried blood-spot levels of C5-acylcarnitines, including isovalerylcarnitine and its isomer, pivaloylcarnitine, which is derived from pivalate-generating antibiotics, has caused many false-positive results. We have developed a method to
S I Goodman et al.
Pediatric research, 14(1), 12-17 (1980-01-01)
When amino acids were infused at a rate of 4 g/kg/day, an infant with hypoglycemia, metabolic acidemia and chronic regurgitation showed hypersarcosinemia and excreted abnormal amounts of sarcosine, isovalerylglycine, isobutyrylglycine, alpha-methylbutyrylglycine, and beta-hydroxyisovaleric, glutaric, alpha-hydroxyglutaric, methylsuccinic, and alpha-hydroxyisobutyric acids in
M H Fries et al.
The Journal of pediatrics, 129(3), 449-452 (1996-09-01)
To assess the effectiveness of glycine and carnitine therapy on isovaleryl conjugate excretion in isovaleric acidemia (IVA). Urinary isovalerylglycine (IVG) and isovalerylcarnitine (IVC) were measured from 12-hour urine specimens collected overnight from an 8-year-old patient with IVA (who had no
C D Capo-chichi et al.
The American journal of clinical nutrition, 69(4), 672-678 (1999-04-10)
Thyroid hormones, riboflavin, riboflavin cofactors, and organic acids were assessed in girls with anorexia nervosa. The objective was to examine the effect of malnutrition and low thyroid hormone concentrations on erythrocyte and plasma riboflavin metabolism and their relation with urinary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service