Skip to Content
Merck
All Photos(1)

Key Documents

SMB01368

Sigma-Aldrich

Octahydrocurcumin

≥95% (HPLC)

Synonym(s):

Hexahydrobisdemethoxycurcumin, Hexahydrocurcuminol, OHC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O6
CAS Number:
Molecular Weight:
376.44
MDL number:
UNSPSC Code:
12352205
UNSPSC Code:
12352200
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

O[C@H](C[C@@H](O)CCc2cc(c(cc2)O)OC)CCc1cc(c(cc1)O)OC

InChI

1S/C21H28O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h5-6,9-12,16-17,22-25H,3-4,7-8,13H2,1-2H3/t16-,17-/m0/s1

InChI key

OELMAFBLFOKZJD-IRXDYDNUSA-N

General description

Octahydrocurcumin (OHC) is an active metabolite of curcumin. OHC has shown anti-inflammatory activity and antioxidant effects on LPS-stimulated inflammation through inhibition of the NF-κB signaling pathway.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing-Feng Xie et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 8856135-8856135 (2021-01-12)
Curcumin (CUR) possesses pronounced anti-inflammatory and antioxidant activities. Generally, the clinical application of CUR is restricted due to its apparent unstability and poor absorption, and the biological activities of CUR may be closely associated with its metabolites. Tetrahydrocurcumin (THC) and
Zhen-Biao Zhang et al.
Frontiers in pharmacology, 9, 1181-1181 (2018-11-06)
Curcumin (CUR), a promising naturally occurring dietary compound, is commonly recognized as the potential anti-inflammatory agent. While the application of CUR was hampered by its low stability and poor systemic bioavailability, it has been suggested that the biological activities of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service