Skip to Content
Merck
All Photos(1)

Documents

NIST885

Refined copper

NIST® SRM® 885

Synonym(s):

Copper

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 200 g

manufacturer/tradename

NIST®

resistivity

1.673 μΩ-cm, 20°C

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

Each unit of SRM 885 consists of a bottle containing 200 g of pins, the material is in the form of pins ~13 mm long, each weighing slightly less than 1 g.

SRM 885_cert

SRM 885_SDS

Application

NIST885 Refined copper standard reference material (SRM) is intended primarily for use in calibration and validation of instrumental methods of analysis used in determinations of sulfur and oxygen in copper and its alloys.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction values of copper matrix.
  • Certified values are metrologically traceable to the SI derived unit of mass fraction expressed as percent (%).  
  • Information values for elements are reported as mass fractions. 
  • The NIST certificate is provided with storage, handling, and use instructions.

Other Notes

Certified for the analytes listed below.
Oxygen (O), Sulfur (S), Silver (Ag), Lead (Pb)

Reference/Informational value is provided for the following analytes.
Antimony (Sb), Arsenic (As), Bismuth (Bi), Iron (Fe), Nickel (Ni), Selenium (Se), Tellurium (Te), Tin (Sn), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service