Skip to Content
Merck

11-0960

Basic Fuchsin

SAJ first grade

Synonym(s):

Basic Violet 14, Fuchsin basic, Magenta, Rosaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H20ClN3
CAS Number:
Molecular Weight:
337.85
MDL number:
UNSPSC Code:
12352106
EC Number:
211-189-6
Beilstein/REAXYS Number:
4166684
Colour Index Number:
42510
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

SAJ first grade

availability

available only in Japan

SMILES string

Cl[H].Cc1cc(ccc1N)\C(c2ccc(N)cc2)=C3\C=CC(=N)C=C3

InChI

1S/C20H19N3.ClH/c1-13-12-16(6-11-19(13)23)20(14-2-7-17(21)8-3-14)15-4-9-18(22)10-5-15;/h2-12,21H,22-23H2,1H3;1H

InChI key

NIKFYOSELWJIOF-UHFFFAOYSA-N

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library