PYR0MATIL6
PyroMAT® Interleukin IL-6 standard
Interleukin-6 standard for use in the Monocyte Activation Test (MAT) for Pyrogen testing
Sign Into View Organizational & Contract Pricing
All Photos(6)
About This Item
Agency
EP Chapter 2.6.30
Quality Level
packaging
pkg of 1 vial
manufacturer/tradename
PyroMAT®
application(s)
pharmaceutical
pyrogen testing
compatibility
configured for PyroMAT®
shipped in
ambient
storage temp.
2-8°C
suitability
pyrogenic bacteria
Related Categories
General description
PYROMAT® IL-6 Control is a recombinant human Interleukin 6 (IL-6) in buffered preservatives, supplied in a lyophilized form. This is used for control reactions in the IL?6 ELISA. The PYROMAT® System is a ready-to-use solution for pyrogen testing, using the Monocyte Activation Test.
Application
The PYROMAT® IL-6 control is to be used with the PYROMAT® System (Monocyte Activation Test for pyrogen detection) for Product Specific Validation to assess that the product does not interfere with the ELISA detection method.
Features and Benefits
- Easy resuspension
- To be used with ELISA kit found in the PYROMAT® system
Components
Recombinant human IL-6 in buffered preservatives, supplied in a lyophilized form
Reconstitution
- Reconstitute the IL-6 Control with endotoxin-free water. The volume of reconstitution can be found on the vial label
- Vortex during 15 min with gentle agitation prior to making dilution
- Aliquot if needed: 20 aliquots can be made from one vial
Recommended products
To be used with PYROMAT® Kit (PYR0MATKIT)
Legal Information
PYROMAT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service