Skip to Content
Merck
All Photos(2)

Key Documents

MABC195

Sigma-Aldrich

Anti-FYVE Antibody, clone 7F11.1

clone 7F11.1, from mouse

Synonym(s):

Zinc finger FYVE domain-containing protein 26, FYVE domain-containing centrosomal protein, FYVE-CENT, Spastizin, FYVE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F11.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZFYVE26(23503)

Related Categories

General description

FYVE, also known as Zinc finger FYVE domain-containing protein 26, or FYVE domain-containing centrosomal protein (FYVE-CENT), or Spastizin, and encoded by the gene ZFYVE26/KIAA0321, is a phosphosphatidylinositol 3-phosphate-binding protein required for the abscission step in cytokinesis. FYVE is recruited to the midbody during cytokinesis and acts as a regulator of abscission. FYVE also be required for efficient homologous recombination DNA double-strand break repair. FYVE is cytoskeleton associated and localized in the cytoplasm in resting cells but at the centrosome during cell division. FYVE is expressed in all cells and tissues. Defects in FYVE may be associated with Spastic paraplegia 15, a form of spastic paraplegia, a neurodegenerative disorder characterized by a slow, gradual, progressive weakness and spasticity of the lower limbs.

Immunogen

GST-tagged recombinant protein corresponding to human FYVE.

Application

Anti-FYVE Antibody, clone 7F11.1 detects level of FYVE & has been published & validated for use in FYVE.
Immunohistochemisty Analysis: A 1:50 dilution from a representative lot detected FYVE in human frontal cortex and thalamus tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Immunohistochemistry in human cerebellum tissue.

Immunohistochemisty Analysis: A 1:50 dilution from a representative lot detected FYVE in human cerebellum tissue.

Target description

~285 kDa calculated. Four isoforms are known to exist at 285 kDa (isoform 1), 283 kDa (isoform 2), 78 kDa (isoform 3), and 89 kDa (isoform 4).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service