Skip to Content
Merck
All Photos(1)

Key Documents

ET12/2-3D0

Millipore

NovaSeptum® GO Tri-clamp Holder, 1-port

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

material

303 stainless steel nut (locking nut and tool)
polysulfone ring(s) (safety ring )

Quality Level

product line

NovaSeptum® GO

feature

autoclavable

parameter

-80-150 °C temp. range (-112-302 °F)
6 bar max. pressure (87 psi) at ±25 °C

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pharma/biopharma processes
pyrogen testing
sterile sampling
sterility testing
viral therapy

storage temp.

room temp

General description

Device configuration: Sterile Sampling connectors
The NovaSeptum® GO TC holder is designed to be used with NovaSeptum® GO sampling units. The holder is also made to fit a TC. For an optimal aseptic solution, use the holder together with a NovAseptic® connector.

Specificity

NovaSeptum® GO sampling holders are designed to be used with a 2 mm diameter needle size that accomodates a broad range of applications. For enhanced protection, each holder can be used with the NovaSeptum® GO Security Tag (ENSTAG) to prevent accidental actuation. A certificate of quality is included in each box.

Application

ET12/2-3D0 (1 shot, 1 unit per box)
Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications

Components

  • Magazine: Glass fiber reinforced Polysulfone (PSU)
  • Safety ring: PSU
  • Locking nut and tool: Stainless steel in compliance with SS2346, ASTM® 303
  • Locking tag: Polyamid/stainless steel
  • Wetted materials: Stainless steel in compliance with 316L EN 1.4435, ASME SA 479/ASTM® A 479, EN 10 272

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service