Skip to Content
Merck
All Photos(1)

Documents

1.19507

Supelco

Sodium standard solution

traceable to SRM from NIST NaNO₃ in H₂O 1000 mg/l Na Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

pH

7 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

format

single component solution

storage temp.

15-25°C

Related Categories

Application



  • Utilization in Bioprinting Research: Sodium standard solutions are used to measure cell proliferation in bioprinting research, aiding in the validation of cell viability and proliferation metrics. This application underscores the importance of precise sodium ion quantification in tissue engineering (Schweinitzer et al., 2024, https://doi.org/10.1088/1748-605X/ad3700).


  • Quantitative Plant Proteomics: Sodium standards are critical for maintaining method linearity in quantitative plant shotgun proteomics, ensuring accurate measurement of protein concentrations. Their application is vital for reproducible quantification in biochemical assays (Leonova et al., 2022, https://doi.org/10.3389/fpls.2022.874761).


  • Evaluation of Mineral Composition: High-purity sodium standards are utilized in the evaluation of mineral compositions, serving as reference points in comparative studies of various biological samples. This usage highlights their role in precise and traceable analytical assessments (Evuen et al., 2022, https://doi.org/10.1038/s41598-022-25204-3).


Analysis Note

Concentration     β (Na): 990 - 1010 mg/l
Determination method: alcalimetric titration.
(traceable to NIST - SRM 723b)
Accuracy of the method: +/- 2 mg

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service