Key Documents
Z188816
Supelco
Black plastic caps
Solid top, white rubber liner, thread for 18-400, pkg of 500 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
black phenolic cap
white rubber liner
feature
closure type black phenolic solid top cap
packaging
pkg of 500 ea
fitting
thread for 18-400
General description
These autoclavable caps are designed to ensure the ultimate in product safety.