SYNF809600
Synthware™ Buchner funnel with perforated plate
600 mL, plate diam. 90 mm
Synonym(s):
Synthware F809600, filtering funnel
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F809600
capacity
600 mL
600 mL
plate diam.
90 mm
Related Categories
General description
With sealed-in perforated glass disc to support filter media. Pressure may be used but should not exceed 15 psi. This design solve the contamination problem with fritted disc and also the blindness problem with ceramic Buchner funnel.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)