CLS6140100
Pyrex® long stem funnel with beaded edge
Synonym(s):
Corning® 6140
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Recommended Products
manufacturer/tradename
Corning 6140-100
stem O.D.
9 mm
top I.D.
100 mm
Related Categories
General description
60° cone angle. Stem length is 150 mm.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)