Skip to Content
Merck
All Photos(1)

Key Documents

630187

Sigma-Aldrich

Diiodomethane-13C,d2

≥99 atom % 13C, ≥98 atom % D, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Methylene iodide-13C,d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CD2I2
CAS Number:
Molecular Weight:
270.84
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

≥99 atom % 13C
≥98 atom % D

Assay

≥99% (CP)

contains

copper as stabilizer

bp

68 °C/11 mmHg (lit.)

mp

6 °C (lit.)

density

3.362 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H][13C]([2H])(I)I

InChI

1S/CH2I2/c2-1-3/h1H2/i1+1D2

InChI key

NZZFYRREKKOMAT-WGVGGRBOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service