Skip to Content
Merck
All Photos(2)

Key Documents

259241

Sigma-Aldrich

2,6-Dichlorophenylacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2CO2H
CAS Number:
Molecular Weight:
205.04
Beilstein:
1952744
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

158-161 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)Cc1c(Cl)cccc1Cl

InChI

1S/C8H6Cl2O2/c9-6-2-1-3-7(10)5(6)4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

SFAILOOQFZNOAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective protonation of samarium enolates by a C2-symmetric chiral diol.
Takeuchi S, et al.
Tetrahedron Asymmetry, 5(9), 1763-1780 (1994)
Ray J Butcher et al.
Acta crystallographica. Section E, Structure reports online, 69(Pt 1), o46-o47 (2013-03-12)
In the title compound, C19H17Cl2N3O2, the amide group is planar and, through N-H⋯O hydrogen bonding to an adjoining mol-ecule, forms dimers of the R2(2)(10) type. As a result of steric repulsion, the amide group is rotated with respect to both
Ye Zhou et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(27), 5404-5418 (2019-05-08)
The absence of functional peripheral myelin protein 22 (PMP22) is associated with shortened lifespan in rodents and severe peripheral nerve myelin abnormalities in several species including humans. Schwann cells and nerves from PMP22 knock-out (KO) mice show deranged cholesterol distribution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service