SML3169
NR162
≥98% (HPLC)
Synonym(s):
4-(Cyclopentylamino)-2-(2,5-dibromo-4-methylbenzylamino)-N-(3-(2-oxooxazolidin-3-yl)propyl)pyrimidine-5-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H30Br2N6O3
Molecular Weight:
610.34
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
NR162 is a potent and selective (per DiscoverX KINOMEScan) inhibitor against the calcium/calmodulin-dependent serine protein kinase CASK (Kd = 22 nM by ITC; IC50 = 80 nM by NanoBRET) with much reduced or little potency against other kinases tested (IC50 = 3.8 μM/TYRO3, 18.2 μM/ERBB3, >20 μM/ABL1, AURKC, RPS6KA6, PIKFYVE, RPS6KA5, LRRK2, STK38L by NanoBRET).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service