CLS430181
Corning® polypropylene beaker
lid only, for 250 mL beaker, sterile, 500/cs
Synonym(s):
Corning container
All Photos(1)
About This Item
Recommended Products
material
natural polypropylene (beaker)
orange high-density polyethylene (snap cap)
sterility
sterile
packaging
pack of 20
case of 500
manufacturer/tradename
Corning 430181
General description
These replacement snap-on polyethylene lids fit on the Corning 250mL containers (CLS430179 and CLS430180) and are certified nonpyrogenic.
Corning flexible polypropylene container with snap-on polyethylene lid serves as a beaker or storage container.
- Container is graduated in both millilitres and ounces
- Certified nonpyrogenic
Legal Information
Corning is a registered trademark of Corning, Inc.