Y0001882
Milbemycin oxime for system suitability
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Milbemycin oxime
About This Item
grade
pharmaceutical primary standard
API family
milbemycin
manufacturer/tradename
EDQM
format
neat
storage temp.
2-8°C
InChI
1S/C32H45NO7.C31H43NO7/c1-6-27-21(4)12-13-31(40-27)17-25-16-24(39-31)11-10-20(3)14-19(2)8-7-9-23-18-37-29-28(33-36)22(5)15-26(30(34)38-25)32(23,29)35;1-18-7-6-8-23-17-36-28-27(32-35)21(4)14-26(31(23,28)34)29(33)37-25-15-24(10-9-19(2)13-18)39-30(16-25)12-11-20(3)22(5)38-30/h7-10,15,19,21,24-27,29,35-36H,6,11-14,16-18H2,1-5H3;6-9,14,18,20,22,24-26,28,34-35H,10-13,15-17H2,1-5H3/b8-7+,20-10+,23-9+,33-28-;7-6+,19-9+,23-8+,32-27-/t19-,21-,24+,25-,26-,27+,29+,31+,32+;18-,20-,22+,24+,25-,26-,28+,30-,31+/m00/s1
InChI key
CKVMAPHTVCTEMM-ALPQRHTBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service