Skip to Content
Merck
All Photos(1)

Key Documents

MABS194

Sigma-Aldrich

Anti-c-K-Ras Antibody, clone 234-4.2

clone 234-4.2, from mouse

Synonym(s):

GTPase Kras, K-Ras 2, Ki-Ras, c-K-ras, c-Ki-ras

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

234-4.2, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (immunogen homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KRAS(3845)

General description

GTPase KRas (c-k-ras; K-ras) is a membrane-bound G protein belonging to the family of Ras GTPases. They are activated by the binding of GTP in response to signals from various receptors including the epidermal growth factor receptor (EGFR). Ras GTPases are regulated by guanine nucleotide exchange factors (GEF) and GTPase activating proteins (GAP) and lie upstream of the MAP kinase pathway which controls a range of cellular processes including proliferation and differentiation. Previous studies have suggested that KRas plays an important role in embryonic development, and it may also contribute to the pathogenesis of colorectal cancer via an EGFR-mediated pathway.

Immunogen

Recombinant protein corresponding to rat c-K-Ras.

Application

Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
This Anti-c-K-Ras Antibody, clone 234-4.2 is validated for use in Western Blotting, IHC for the detection of c-K-Ras.
Western Blot Analysis: A representative lot detected c-K-Ras in A431 cell lysate.

Western Blot Analysis: A representative lot from an independent laboratory detected c-K-Ras in primary human
lymphoid leukemia cell lysates, and in normal and HA-dynamin K44A mutant HeLa cell lysates (Omerovic, J., et al. (2008). Oncogene. 27(19):2754-2762.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected c-K-Ras in non-small cell lung cancer cells (Volm, M., et al. (2002). Clin Cancer Res. 8(6):1843-1848.).

Quality

Evaluated by Western Blot in SW-480 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected c-K-Ras in 10 µg of SW-480 cell lysate.

Target description

~22 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
SW-480 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaine Lai-Han Leung et al.
International journal of cancer, 145(5), 1334-1345 (2019-02-21)
Oncogenic KRAS is considered a promising target for anti-cancer therapy. However, direct pharmacological strategies targeting KRAS-driven cancers remained unavailable. The prenyl-binding protein PDEδ, a transporter of KRAS, has been identified as a potential target for pharmacological inhibitor by selectively binding
Enkhtsetseg Munkhbaatar et al.
Nature communications, 11(1), 4527-4527 (2020-09-12)
Evasion of programmed cell death represents a critical form of oncogene addiction in cancer cells. Understanding the molecular mechanisms underpinning cancer cell survival despite the oncogenic stress could provide a molecular basis for potential therapeutic interventions. Here we explore the
Almas Yaqoob et al.
PloS one, 15(5), e0231948-e0231948 (2020-05-06)
In our search for bioactive mushrooms native to British Columbia, we determined that the ethanol extracts from fruiting bodies of the terrestrial polypore Albatrellus flettii had potent anti-cell viability activity. Using bioassay-guided fractionation, mass spectrometry and nuclear magnetic resonance, we
Caterina Bartolacci et al.
Nature communications, 13(1), 4327-4327 (2022-07-27)
Mutant KRAS (KM), the most common oncogene in lung cancer (LC), regulates fatty acid (FA) metabolism. However, the role of FA in LC tumorigenesis is still not sufficiently characterized. Here, we show that KMLC has a specific lipid profile, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service