Skip to Content
Merck
All Photos(1)

Key Documents

AB5404

Sigma-Aldrich

Anti-Neurturin Antibody

serum, Chemicon®

Synonym(s):

Anti-Anti-NTN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NRTN(4902)

Specificity

Neurturin. No cross reactivity with GDNF.

Immunogen

Recombinant human Neurturin.

Application

Anti-Neurturin Antibody detects level of Neurturin & has been published & validated for use in IH.
Immunohistochemistry: 1:1,000-1:2,000

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Physical form

Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50 μL of sterile distilled water. Centrifuge if necessary to remove any residue.

Storage and Stability

Maintain lyophilized material at -20°C to -70°C for up to 6 months. After reconstitution maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suddhasil Mookherjee et al.
Human molecular genetics, 24(22), 6446-6458 (2015-09-12)
Retinal neurodegenerative diseases are especially attractive targets for gene replacement therapy, which appears to be clinically effective for several monogenic diseases. X-linked forms of retinitis pigmentosa (XLRP) are relatively severe blinding disorders, resulting from progressive photoreceptor dysfunction primarily caused by
James F Striebel et al.
Acta neuropathologica communications, 9(1), 17-17 (2021-01-30)
Accumulation of misfolded host proteins is central to neuropathogenesis of numerous human brain diseases including prion and prion-like diseases. Neurons of retina are also affected by these diseases. Previously, our group and others found that prion-induced retinal damage to photoreceptor
E M Boggio et al.
Neuroscience, 324, 496-508 (2016-03-24)
The Forkead Box G1 (FOXG1 in humans, Foxg1 in mice) gene encodes for a DNA-binding transcription factor, essential for the development of the telencephalon in mammalian forebrain. Mutations in FOXG1 have been reported to be involved in the onset of
Xiao Li et al.
The FEBS journal, 290(17), 4356-4370 (2023-04-26)
The endoplasmic reticulum membrane protein complex (EMC) plays a critical role in the synthesis of multipass membrane proteins. Genetic studies indicated that mutations in EMC1 gene were associated with retinal degeneration diseases; however, the role of EMC1 in photoreceptor has
Guobao Li et al.
Frontiers in molecular neuroscience, 12, 126-126 (2019-06-11)
The centrosomal protein γ-tubulin complex protein 3 (Tubgcp3/GCP3) is required for the assembly of γ-tubulin small complexes (γ-TuSCs) and γ-tubulin ring complexes (γ-TuRCs), which play critical roles in mitotic spindle formation during mitosis. However, its function in vertebrate embryonic development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service