Key Documents
Z183954
Sigma-Aldrich
Aldrich® large-capacity cold trap
Inside finger for traps with 50mm O-ring
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.
General description
With removable cold-fingers and a choice of 55/50 joint or 50 mm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)