WHA9916-1304
Whatman® Uniflo® 13 syringe filters
polyethersulfone membrane, sterile, pore size 0.45 μm, pack of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polyethersulfone membrane
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
9916-1304
diam.
13 mm
pore size
0.45 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- 13 or 25 mm diameters
- 0.2 µm, 0.22 µm or 0.45 µm pore sizes
- Sterile or non-sterile options
- Individual printing on the filter for easy identification
- Bench top space saving packaging
Other Notes
Suitable for volumes of 10 mL or less
Legal Information
UNIFLO is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service