WHA10371025
Whatman® standard glass microfiber extraction thimbles
603G glass fiber thimble 28 x 60 mm - thickness 1.5 mm, 25/pk
Synonym(s):
Z612480, extraction thimbles, glass fiber filters, glass fibre filters
About This Item
Recommended Products
description
Grade 603 G (Glass Fiber with inorganic binder)
packaging
pack of 100
manufacturer/tradename
Whatman 10371025
Whatman Article No. 28414813 (US reference)
I.D. × L
28 mm × 60 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Cytiva′s standard glass Soxhlet thimble range
Whatman standard glass extraction thimbles are available in a range of dimensions and wall thicknesses to suit your application:
- Twelve internal diameter options
- Twelve thimble length options
Features and Benefits
- Resistant to temperatures up to 500°C (932°F)
- Manufactured with walls as thin as 1–1.5 mm
- Analysis of hot and acidic gases, including emission control and smoke stack gas monitoring
- Soxhlet extraction
- Analysis of pesticide residues
- Determination of oil/fat content of foods
- Analysis of oil and grease in solid wastes
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service