Skip to Content
Merck
All Photos(2)

Key Documents

DWKBGE218P

Azlon Square Bottles

graduated, cap, Plug, translucent high-density polyethylene, capacity 50 mL

Synonym(s):

BGE218P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

material

translucent high-density polyethylene

sterility

non-sterile

feature

cap Plug
graduated

packaging

pkg of 10 ea

manufacturer/tradename

Azlon

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for storage of liquid and powder samples

Features and Benefits

Space saving square design

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service