MilliporeSigma
  • Biologically active acylglycerides from the berries of saw-palmetto (Serenoa repens).
Biologically active acylglycerides from the berries of saw-palmetto (Serenoa repens).
Journal of natural products (1997-04-01)
H Shimada, V E Tyler, J L McLaughlin
ABSTRACT
Brine shrimp lethality-directed fractionation of the 95% EtOH extract of the powdered, dried berries of Serenoa repens (Bart.) Small (saw-palmetto) (Palmae) led to the isolation of two monoacylglycerides, 1-monolaurin (1) and 1-monomyristin (2). Compounds 1 and 2 showed moderate biological activities in the brine shrimp lethality test and against renal (A-498) and pancreatic (PACA-2) human tumor cells; borderline cytotoxicity was exhibited against human prostatic (PC-3) cells. The fruits and extracts of saw-palmetto are taken orally as an herbal medicine to prevent prostatic hyperplasias.
Sigma-Aldrich
1-Lauroyl-rac-glycerol, ≥99%