Merck
  • Assessment of expressions of Bcl-XL, b-FGF, Bmp-2, Caspase-3, PDGFR-α, Smad1 and TGF-β1 genes in a rat model of lung ischemia/reperfusion.
Assessment of expressions of Bcl-XL, b-FGF, Bmp-2, Caspase-3, PDGFR-α, Smad1 and TGF-β1 genes in a rat model of lung ischemia/reperfusion.
Iranian journal of basic medical sciences (2016-04-16)
Hasan Şimşek, Şeniz Demiryürek, Tuncer Demir, Hüsne Didem Atabay, Ali Osman Çeribasi, Recep Bayraktar, Davut Sinan Kaplan, Serdar Öztuzcu, Beyhan Cengiz
ABSTRACT
Ischemia is described as organs and tissues are destitute of oxygen due to decreased arterial or venous blood flow. Many mechanisms play role in cell death happened as a consequence of a new blood flow is needed for both cell regeneration and to clean toxic metabolites during ischemia and later. Lung damage induced by ischemia/reperfusion (I/R) is a frequent problem in lung transplantation. Apoptosis (programmed cell death) is known as cell suicide, and plays a key role in embryonic developmental and in maintain adult tissue's life. It is investigated expressions of Smad1, Bmp-2, Bcl-XL, b-FGF, Caspase-3, TGF-β1, PDGFR-α genes for molecular changes in lung tissues, after I/R is formed, in this study. For this, we included 40 Wistar albino rats to this study and divided 4 groups (n=10). The Groups were determined as Control (C), Group 1= 1 hr ischemia (I), Group 2= 1 hr ischemia+2 hr reperfusion (I+2R), Group 3= 1 hr ischemia+4 hr reperfusion (I+4R). Besides, molecular analysis and histopathologic examinations of tissues were performed, and the results were evaluated by normalization and statistics analysis. We have found a significant increase in expression of Bcl-XL (P=0.046) and Caspase-3 (P=0.026) genes of group 1, and it was not monitored any significant difference in Group 2 and Group 3. In all groups, the changes in b-FGF (P=0.087), Bmp-2 (P=0.457), TGF-β1 (P=0.201) and PDGFR-α (P=0.116) were not significant compared to control group. We did not see any mRNA expression of Smad1 gene in all groups include control. These findings suggest that I/R injury may trigger apoptotic mechanism in lung.
Product Description
Roche
High Pure RNA Tissue Kit, sufficient for 50 isolation(s), suitable for RT-PCR, suitable for Northern blotting