Key Documents
58817
Rheodyne® Model 7060 Selection Valve
About This Item
UNSPSC Code:
41115854
feature
number of ports 7
positions 6
parameter
5000 psi max. pressure (350 bar) (factory set)
General description
For column or mobile phase selection. Instead of frequently connecting and disconnecting columns to your system, connect the injector outlet to the common center port of a Model 7060 valve, and connect up to 6 columns to the peripheral ports. (We recommend allowing one port for bypass/flushing operations.) The common port can be sequentially connected to each of the 6 peripheral ports by manually rotating the handle in either direction. Connect the column outlets to a second Model 7060 valve to direct the flow from the columns to the detector. Low dispersion 0.41 mm/0.016 in. I.D. passages and an internal volume of less than 2 μL ensure minimal extra-column band broadening.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)