Documents
58626
Supelco
Tee
zero dead volume, for connecting 1/16 in., bore size 0.25 mm
Synonym(s):
Valco® Unions
About This Item
UNSPSC Code:
41121815
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® ZT1C
bore size
0.25 mm
fitting
10-32 female Valco for 1/16 in. O.D. tubing (fittings included)
compatibility
for connecting 1/16 in.
General description
Stainless steel, unless noted otherwise. Nuts and ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)