Documents
57653
Upchurch PEEK LiteTouch® Fitting
double-winged natural PEEK nut
About This Item
UNSPSC Code:
41115700
material
double-winged natural PEEK nut
description
litetouch fitting
packaging
pkg of 10 ea
fitting
1/4-28 male unit for 1/8 in. O.D. tubing (requires LiteTouch ferrule)
General description
Unique design prevents twisting of PEEK tubing. The fitting consists of a special PEEK ferrule, a stainless steel ring, and a PEEK or stainless steel nut for 1/16 in. tubing. The fitting will not leak at pressures to 5000 psi/340 bar with the PEEK nut, 2500 psi/175 bar with the stainless steel nut finger-tightened or 7000 psi/480 bar with wrench tightening. After wrench-tightening the stainless steel nut, subsequent finger-tightening will hold to 5000 psi. Compatible with all manufacturers′ stainless steel nuts. Order nuts and ferrules separately.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
LiteTouch is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Pedram Madadkar et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1055-1056, 158-164 (2017-05-10)
Cation exchange (CEX) chromatography is widely used for large-scale separation of monoclonal antibody (mAb) aggregates. The aggregates bind more strongly to CEX media and hence elute after the monomeric mAb in a salt gradient. However, monomer-aggregate resolution that is typically