Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56666-U

Supelco

Micro-Mixer Static Mixers (2 - 25 μL)

housing, Binary

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

material

stainless steel

Looking for similar products? Visit Product Comparison Guide

General description

Micro-Mixer of Binary input housing is made of stainless steel, and is a part of ASI Static Mixers, with a highly efficient cross-flow shearing mechanism, which produces vortex mixing over a wide range of flow rates. Binary input housing is used to combine two flow paths into one, such as in post-column or gradient mixing applications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service