Key Documents
55082
Upchurch Tubing Adapter
About This Item
UNSPSC Code:
41115700
material
Tefzel® adapter (one-piece design)
packaging
pkg of 1 ea
fitting
female Luer
10-32 male
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)