Skip to Content
MilliporeSigma
All Photos(1)

Documents

52006

Supelco

5-Geranyloxy-7-methoxycoumarin

analytical standard

Synonym(s):

5-Geranoxy-7-methoxycoumarin, 5-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-7-methoxy-2H-1-benzopyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O4
CAS Number:
Molecular Weight:
328.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

O=C1OC2=CC(OC)=CC(OC/C=C(C)/CCC=C(C)C)=C2C=C1

InChI

1S/C20H24O4/c1-14(2)6-5-7-15(3)10-11-23-18-12-16(22-4)13-19-17(18)8-9-20(21)24-19/h6,8-10,12-13H,5,7,11H2,1-4H3/b15-10+

InChI key

WXUOSNJWDJOHGW-XNTDXEJSSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Arigò et al.
Journal of chromatography. A, 1649, 462183-462183 (2021-05-27)
In the European Union, cosmetic products are regulated by the Cosmetics Regulation (EC) No. 1223/2009. The Analytical Working Group of the International Fragrance Association (IFRA) suggested that a new sensitive analytical method is needed to determine psoralens in finished cosmetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service