Skip to Content
MilliporeSigma
All Photos(8)

Documents

ZRB2777

Sigma-Aldrich

Anti-CD41/ITGA2B Antibody, clone 1F9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CD41), GPIIb, GPalpha IIb, Integrin alpha-IIb, Platelet membrane glycoprotein IIb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1F9
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 114 kDa
observed mol wt ~115 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... ITGA2B(3674)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F9 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects CD41/Integrin 2b. It targets an epitope within 21 amino acids from the extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the extracellular domain of human CD41/Integrin 2b.

Application

Quality Control Testing

Evaluated by Western Blotting in Human spleen tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD41/ITGA2B in Human spleen tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CD41/ITGA2B in Human liver tissue lysate.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected CD41/ITGA2B in one million Human peripheral blood mononuclear cells (PBMC).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD41/ITGA2B in A549 cells.

Affinity Binding Assay: A representative lot of this antibody bound CD41/ITGA2B peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CD41/ITGA2B in Human bone marrow tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Integrin alpha-IIb (UniProt: P08514; also known as GPalpha IIb, GPIIb, Platelet membrane glycoprotein IIb, CD41) is encoded by the ITGA2B (also known as GP2B, ITGAB) gene (Gene ID: 3674) in human. CD41/ITGA2B is a single-pass type I membrane glycoprotein of the integrin chain family. It is synthesized with a signal peptide (aa 1-31), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 32-993), a transmembrane domain (aa 994-1019), and a short cytoplasmic domain (aa 1020-1039). Along with β3 it serves as a receptor of fibronectin, fibrinogen, plasminogen, prothrombin, thrombospondin and vitronectin. It recognizes the sequence R-G-D in a wide array of ligands. Following activation, integrin IIb/β3 brings about platelet/platelet interaction through binding of soluble fibrinogen. This step leads to rapid platelet aggregation, which physically plugs ruptured endothelial cell surface. Mutations in ITGA2B gene have been linked to Glanzmann thrombasthenia 1 (GT1), a disorder characterized by failure of platelet aggregation, absent or diminished clot retraction, and mucocutaneous bleeding of mild-to-moderate severity. Three isoforms of CD41/ITGA2B have been described that are produced by alternative splicing. Isoform 1 and 2 have been identified in platelets and megakaryocytes, but not in reticulocytes or in Jurkat and U-937 cell lines. Isoform 3 is expressed by leukemia, prostate adenocarcinoma, and melanoma cells but not by platelets or normal prostate or breast epithelial cells. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: D′Andrea, G., et al. (2002). Thromb. Haemost. 87(6); 1034-1042; Bray, PF., et al. (1990). J. Biol. Chem. 265(17); 9587-9590).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service