Key Documents
Z768634
Nalgene® square graduated media bottle
with cap, PET, sterile, capacity 125 mL
Synonym(s):
media storage bottle, square plastic bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
PET
square bottle
Joint
Joints threaded neck (38-430)
sterility
sterile
feature
graduations
with cap
packaging
pack of 96 ea (4 x pack of 24)
manufacturer/tradename
Nalgene 842040-0125
capacity
125 mL
Features and Benefits
  • Closure with gas barrier properties that enables storage of liquid media, buffers, and sera
  • The bottles are light-weight, sterile and shatter-resistant
  • Leakproof, transparent, and space-saving square design
  • Polyethylene terephthalate with closure made of high-density polyethylene
  • Graduated and possesses neck ring that can be used with tamper evident shrink sleeves
  • Gamma irradiated- sterility assurance level (SAL) of 10-6
  • Non-cytotoxic and non-pyrogenic
  • Lower permeability towards CO2 and O2
  • Bottle and closure material are Animal Derived Component Free (ADCF)
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)