Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743436

Laser Cryo-Tags® sidewall

yellow, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels

Synonym(s):

cryogenic plate labels, freezer labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103032
NACRES:
NB.35

packaging

pkg of 3120 labels 20 sheets of 156 labels

manufacturer/tradename

Diversified Biotech SIDE-1500

technique(s)

cryopreservation: suitable

size

1.50 in. × 0.25 in.

color

yellow

Looking for similar products? Visit Product Comparison Guide

General description

Sidewall Cryo-Tags for laser printers withstand both long term cryogenic storage conditions and conventional freezer storage.  These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic multiwell plates.  Cryo-Tags also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100°C and dry heat up to 150°C.

Legal Information

D-230 is a registered trademark of Texaco

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service