Key Documents
Z742003
Low Profile Roller Double Platform Stack
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
material
acrylic platform
aluminum support rod
manufacturer/tradename
IBI ACCRL0002
W × D × H
18 in. × 16 in. × 18 3/4 in.
General description
Product contains two each of the raised platforms that will accommodate two additional Low Profile Roller. This raised platform is designed to fit inside an incubator to allow placement of three Low Profile rollers inside.
Constructed of cast acrylic and aluminum for years of service inside high humidity, CO2 incubators without discoloring or rusting. Platforms bolt together for secure placement of Low Profile rollers.
Features and Benefits
  • Allows placement of two additional rollers inside incubator
  • Constructed of cast acrylic and aluminum
  • Save lab counter space by stacking three rollers.
  • Platforms bolt together for secure placement of Low Profile rollers
Choose from one of the most recent versions:
Certificates of Analysis (COA)