Key Documents
Z740482
MICROGARD 2500 coveralls for protection from saturating liquid spray
size L
About This Item
UNSPSC Code:
46181503
NACRES:
NB.35
size
L
manufacturer/tradename
Arco 1724205
General description
MICROGARD 2500 STANDARD is designed to protect from saturating liquid spray and particulates, the fabric′s physical strength and flexibility ensure you are protected yet comfortable.
Features:
  • 3 piece hood
  • Ultrasonically welded seams
  • Finger loops
  • 2-way front zip with resealable storm flap
  • Elasticated hood, waist, wrists and ankles
Approved to:
EN14605 Type 4 Spray-tight
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-1 Anti-static
Legal Information
Microgard is a trademark of Microgard Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)