Documents
Z362735
Sigma-Aldrich
Nalgene® polycarbonate Erlenmeyer flask
capacity 250 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
About This Item
UNSPSC Code:
41121804
eCl@ss:
32040501
NACRES:
NB.43
material
flat bottom
polycarbonate
wide-neck
Joint
joint stopper6
sterility
non-sterile
feature
closure type Stoppers
without cap
packaging
case of 12
pack of 6
manufacturer/tradename
Nalgene 4103-0250
capacity
250 mL
General description
Autoclavable; however, repeated autoclaving may cause some loss of mechanical strength.
Other Notes
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries