Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3188

Sigma-Aldrich

WISP-3 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCN6, WNT-1 inducible signaling pathway protein-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.2-0.3 μg/mL ED50

mol wt

36.8 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WISP3(8838)

General description

WISP-3 is a member of the CCN family of secreted cysteine rich regulatory proteins. It is predominantly expressed in the adult kidney, testis and fetal kidney and weaker expression in placenta, ovary, prostate, small intestine, and skeletally-derived cells. WISP-3 is required for normal postnatal skeletal growth and cartilage homeostasis. Human WISP-3 is a 36.8 kDa protein containing 331 amino acid residues. It is composed of four distinct structural domains (modules); the IGF binding protein (IGFBP) domain, the von Willebrand Factor C (VWFC) domain, the thrombospondin type-1 repeat (TSP type-1) domain, and a C-terminal cystine knot-like (CTCK) domain.

Biochem/physiol Actions

WISP-3 is a member of the CCN family of secreted cysteine rich regulatory proteins. It is predominantly expressed in the adult kidney, testis and fetal kidney and weaker expression in placenta, ovary, prostate, small intestine, and skeletally-derived cells. WISP-3 is required for normal postnatal skeletal growth and cartilage homeostasis. Human WISP-3 is a 36.8 kDa protein containing 331 amino acid residues. It is composed of four distinct structural domains (modules); the IGF binding protein (IGFBP) domain, the von Willebrand Factor C (VWFC) domain, the thrombospondin type-1 repeat (TSP type-1) domain, and a C-terminal cystine knot-like (CTCK) domain.

Sequence

TGPLDTTPEG RPGEVSDAPQ RKQFCHWPCK CPQQKPRCPP GVSLVRDGCG CCKICAKQPG EICNEADLCD PHKGLYCDYS VDRPRYETGV CAYLVAVGCE FNQVHYHNGQ VFQPNPLFSC LCVSGAIGCT PLFIPKLAGS HCSGAKGGKK SDQSNCSLEP LLQQLSTSYK TMPAYRNLPL IWKKKCLVQA TKWTPCSRTC GMGISNRVTN ENSNCEMRKE KRLCYIQPCD SNILKTIKIP KGKTCQPTFQ LSKAEKFVFS GCSSTQSYKP TFCGICLDKR CCIPNKSKMI TIQFDCPNEG SFKWKMLWIT SCVCQRNCRE PGDIFSELKI L

Physical form

Lyophilized from 10 mM Acetic Acid.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 10 mM Acetic Acid to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Hurvitz et al.
Nature genetics, 23(1), 94-98 (1999-09-02)
Members of the CCN (for CTGF, cyr61/cef10, nov) gene family encode cysteine-rich secreted proteins with roles in cell growth and differentiation. Cell-specific and tissue-specific differences in the expression and function of different CCN family members suggest they have non-redundant roles.
D Pennica et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(25), 14717-14722 (1998-12-09)
Wnt family members are critical to many developmental processes, and components of the Wnt signaling pathway have been linked to tumorigenesis in familial and sporadic colon carcinomas. Here we report the identification of two genes, WISP-1 and WISP-2, that are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service